Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSBN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15772420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15772420 20 μL
NBP157724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15772420 Supplier Novus Biologicals Supplier No. NBP15772420UL

Rabbit Polyclonal Antibody

RSBN1 Polyclonal specifically detects RSBN1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen RSBN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q5VWQ0
Gene Alias DKFZp781E21150, FLJ11220, ROSBIN, round spermatid basic protein 1, RP11-324J2.1
Gene Symbols RSBN1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RSBN1 (round spermatid basic protein 1) The peptide sequence was selected from the N terminal of RSBN1)(50ug). Peptide sequence GGAVGPFKCVFVGEMAAQVGAVRVVRAVAAQEEPDKEGKEKPHAGVSPRG.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54665
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.