Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSF1 Antibody (C.15200041), Novus Biologicals™
SDP

Catalog No. NBP259285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259285 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259285 Supplier Novus Biologicals Supplier No. NBP259285
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RSF1 Monoclonal specifically detects RSF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen RSF1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone C.15200041
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 0.3 ug
Formulation PBS with 0.05% Sodium Azide
Gene Alias HBV pX associated protein-8, HBXAPp325 subunit of RSF chromatin-remodeling complex, hepatitis B virus x associated protein, Hepatitis B virus X-associated protein, p325, remodeling and spacing factor 1, Rsf-1, XAP8HBV pX-associated protein 8
Gene Symbols RSF1
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51773
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.