Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSK1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357767 Supplier Novus Biologicals Supplier No. NBP176620MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSK1 Polyclonal antibody specifically detects RSK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen RSK1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human RPS6K1. The immunogen is located within amino acids 600 - 650 of RPS6K1. Amino Acid Squence: ANGPSDTPEEILTR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6195
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.