Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSK1, p Thr573 Antibody, Novus Biologicals™
SDP

Catalog No. NB176958 Shop All R&D Systems Products
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176958 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. NB176958 Supplier Novus Biologicals missing translation for 'supplierNo' NBP313271
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Rabbit Polyclonal Antibody

RSK1 Polyclonal antibody specifically detects RSK1 in Human samples. It is validated for Western Blot

Specifications

Antigen RSK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human RSK1 (phospho Thr573). The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6195
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.