Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSK2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP11929505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11929505 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11929505 Supplier Novus Biologicals Supplier No. NBP1192950.05MG

Rabbit Polyclonal Antibody

RSK2 Polyclonal specifically detects RSK2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RSK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, pH 7.2, containing 50% glycerol. with 0.09% Sodium Azide
Gene Alias 90 kDa ribosomal protein S6 kinase 3, CLS, Coffin-Lowry syndrome, EC 2.7.11, EC 2.7.11.1, HU-3, Insulin-stimulated protein kinase 1, ISPK1, ISPK-1, MAP kinase-activated protein kinase 1b, MAPK-activated protein kinase 1b, MAPKAP kinase 1b, MAPKAPK-1b, MAPKAPK1B, mental retardation, X-linked 19, MRX19, p90-RSK2, p90RSK3, pp90RSK2, ribosomal protein S6 kinase alpha-3, ribosomal protein S6 kinase, 90kD, polypeptide 3, ribosomal protein S6 kinase, 90kDa, polypeptide 3, Ribosomal S6 kinase 2, RSK, RSK-2, RSK2p90-RSK 3, S6K-alpha3, S6K-alpha-3
Gene Symbols RPS6KA3
Host Species Rabbit
Immunogen Synthetic peptide derived from the sequence of mouse RSK2, conjugated to KLH
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6197
Test Specificity p90/RSK2
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.