Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSL1D1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB045807 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045807 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045807 Supplier Novus Biologicals Supplier No. NBP297626AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSL1D1 Polyclonal specifically detects RSL1D1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RSL1D1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CATX11, CATX-11, Cellular senescence-inhibited gene protein, CSIGMGC142259, DKFZP564M182, L12, MGC138433, PBK1cellular senescence inhibited gene protein, Protein PBK1, ribosomal L1 domain containing 1, ribosomal L1 domain-containing protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RSL1D1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26156
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.