Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV/BRSV Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10093585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093585 0.1 mg
NB017933 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10093585 Supplier Novus Biologicals Supplier No. NB10093585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSV/BRSV Polyclonal specifically detects RSV/BRSV in Virus samples. It is validated for Western Blot.

Specifications

Antigen RSV/BRSV
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry
Formulation PBS with 0.02% Sodium Azide
Gene Alias Bovine Respiratory Syncytial Virus BRSV, Bovine RSV
Host Species Rabbit
Immunogen Residues 201-211 [KELLPKVNNHDC] of the 63 kDa BRSV F protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with Respiratory Syncytial Virus (RSV) Fusion Protein of the RB94 strain in bovine samples.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.