Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV/BRSV Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB209674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209674 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209674 Supplier Novus Biologicals Supplier No. NB10093585C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RSV/BRSV Polyclonal antibody specifically detects RSV/BRSV in Virus samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen RSV/BRSV
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Bovine Respiratory Syncytial Virus BRSV, Bovine RSV
Host Species Rabbit
Immunogen Residues 201-211 [KELLPKVNNHDC] of the 63 kDa BRSV F protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.