Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB151070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151070 Supplier Novus Biologicals Supplier No. NBP275191AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RSV Fusion Protein Monoclonal specifically detects RSV Fusion Protein in Virus samples. It is validated for ELISA.

Specifications

Antigen RSV Fusion Protein
Applications ELISA
Classification Monoclonal
Clone RSHZ19 (Felvizumab)
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.