Learn More
RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), DyLight 488, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | RSV Fusion Protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | RSHZ19 (Felvizumab) |
| Conjugate | DyLight 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Human |
| Immunogen | This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.