Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSV Fusion Protein Antibody (RSHZ19 (Felvizumab)), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343048 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343048 Supplier Novus Biologicals Supplier No. NBP275190MFV610
Add to Cart
Add to Cart

Human Monoclonal Antibody

RSV Fusion Protein Monoclonal antibody specifically detects RSV Fusion Protein in Virus samples. It is validated for ELISA, Functional Assay

Specifications

Antigen RSV Fusion Protein
Applications ELISA, Functional Assay
Classification Monoclonal
Clone RSHZ19 (Felvizumab)
Conjugate mFluor Violet 610 SE
Dilution ELISA, Functional
Formulation 50mM Sodium Borate
Gene Alias Human respiratory syncytial virus, RSV, RSV fusion protein, Rsvgp08
Host Species Human
Immunogen This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.