Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTEL1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222360SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222360SS 0.025 mL
NBP222360 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222360SS Supplier Novus Biologicals Supplier No. NBP222360SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

RTEL1 Polyclonal specifically detects RTEL1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen RTEL1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.26 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:250 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:300
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NZ71
Gene Alias C20orf41, regulator of telomere elongation helicase 1, regulator of telomere length
Gene Symbols RTEL1
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human RTEL1 protein (within residues 800-1000). [Swiss-Prot Q9NZ71]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 51750
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.