Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTF1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB600278AFB Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600278AFB 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600278AFB Supplier Novus Biologicals Supplier No. NB600278AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTF1 Polyclonal specifically detects RTF1 in Human samples. It is validated for Western Blot.

Specifications

Antigen RTF1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KIAA0252GTL7, ortholog of mouse gene trap locus 7, RNA polymerase-associated protein RTF1 homolog, Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae)
Gene Symbols RTF1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 620 and 670 of human Restores TBP (TATA box-binding protein) function-1 using the numbering given in entry EAW92492.1 (GeneID 23168).
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23168
Test Specificity Specific for human Rtf1 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.