Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN1-A/NSP Antibody (MON162), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP197677X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197677X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197677X Supplier Novus Biologicals Supplier No. NBP197677AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen RTN1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MON162
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias Neuroendocrine-specific proteinNSPMGC133250, reticulon 1, reticulon-1
Gene Symbols RTN1
Host Species Mouse
Immunogen MON-162 is a Mouse monoclonal IgG1 antibody derived by fusion of Mouse myeloma cells with spleen cells from a Mouse immunized with a partially purified bacterially expressed Reticulon-1A (NSP-A) hybrid protein (beta-GAL-NSP-A 6-776). The epitope of MON-16
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6252
Test Specificity MON-162 exclusively recognizes the 135 kD Reticulon-1A protein in immunoblots of NCI-H82 and other SCLC cell lines, and stains normal and pathological neural and neuroendocrine tissues.
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.