Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN1-A/NSP Antibody (RNL-4), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N197673PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N197673PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N197673PEC5 Supplier Novus Biologicals Supplier No. NBP197673PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Human, Rat, Porcine samples. It is validated for Flow Cytometry.

Specifications

Antigen RTN1-A/NSP
Applications Flow Cytometry
Classification Monoclonal
Clone RNL-4
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Symbols RTN1
Host Species Mouse
Immunogen RNL-4 is a Mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells, with spleen cells from a BALB/c Mouse immunized with a synthetic peptide encompassing the first 20 amino acids on the N-terminal of Reticulon-1C (NSP-C).
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6252.0
Test Specificity RNL-4 reacts with peripheral nerves and ganglia of various tissues and cross-reacts with smooth muscle cells and myoepithelium. In the central nervous system it reacts with the neurohypophysis and pars intermedia of the pituitary gland, and a weak diffuse
Target Species Human, Rat, Rat (Negative)
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.