Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN4/NOGO Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7088224 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6555 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6555 Supplier Proteintech Group Inc Supplier No. 107401AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NogoA is a member of a family of integral membrane proteins termed reticulons that are thought to be involved in numerous disorders including neurodegenerative diseases. Reticulon proteins are known to regulate many cellular processes and interact with multiple proteins and receptors such as BACE. NogoA was initially identified as a myelin-associated neurite outgrowth inhibitor. It is highly expressed in oligodendrocytes in the white matter of the CNS, blocking its activity with antibodies or other factors results in improved axon regrowth and functional recovery in experimental CNS lesion models. NogoA has also been suggested to play a role in neurodegenerative diseases such as Amyotrophic lateral sclerosis, in which case NogoA is found at elevated levels in postmortem muscular samples, and multiple sclerosis (MS), in which case autoantibodies to NogoA have been found in serum and cerebrospinal fluid in MS patients.

Specifications

Antigen RTN4/NOGO
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RTN4
Gene Accession No. Q99P72, Q9NQC3
Gene Alias ASY, Foocen, KIAA0886, Nbla00271, Nbla10545, Neurite outgrowth inhibitor, NI220/250, NOGO, NOGO A, Nogo B, Nogo C, Nogo protein, NOGOA, NOGOC, NSP, NSP CL, reticulon 4, Reticulon 5, RTN X, RTN4, RTN4 A, RTN4 B1, RTN4 B2, RTN4 C, SP1507
Gene Symbols RTN4
Host Species Rabbit
Immunogen RTN4/NOGO Fusion Protein Ag1164
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57142, 68585
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.