Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTVP-1/GLIPR1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB058965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB058965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB058965 Supplier Novus Biologicals Supplier No. NBP299206C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTVP-1/GLIPR1 Polyclonal specifically detects RTVP-1/GLIPR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RTVP-1/GLIPR1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GLI pathogenesis-related 1, GLI pathogenesis-related 1 (glioma), GliPR, GliPR 1, GLIPRglioma pathogenesis-related protein 1, Protein RTVP-1, related to testis-specific, vespid, and pathogenesis proteins 1, RTVP1CRISP7, testes-specific vespid and pathogenesis protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RTVP-1/GLIPR1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11010
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.