Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTVP-1/GLIPR1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB341422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341422 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341422 Supplier Novus Biologicals Supplier No. NBP282032MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RTVP-1/GLIPR1 Polyclonal antibody specifically detects RTVP-1/GLIPR1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen RTVP-1/GLIPR1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GLI pathogenesis-related 1, GLI pathogenesis-related 1 (glioma), GliPR, GliPR 1, GLIPRglioma pathogenesis-related protein 1, Protein RTVP-1, related to testis-specific, vespid, and pathogenesis proteins 1, RTVP1CRISP7, testes-specific vespid and pathogenesis protein 1
Host Species Rabbit
Immunogen GLIPR1 antibody was raised against an 18 amino acid peptide near the center of human GLIPR1. The immunogen is located within amino acids 60 - 110 of GLIPR1. Amino Acid Squence: QFSHNTRLKPPHKLHPN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 11010
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.