Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rtx™-5 Capillary Columns, 0.53 mm I.D.
SDP

Catalog No. 06610662 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
15 m
30 m
60 m
Film Thickness:
0.1 μm
0.25 μm
1 μm
3 μm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Film Thickness
06-610-662 60 m 1 μm
06-610-647 15 m 0.1 μm
06-610-649 30 m 0.1 μm
06-610-652 15 m 0.25 μm
06-610-655 30 m 0.25 μm
06-610-657 60 m 0.25 μm
06-610-666 15 m 3 μm
06-610-669 60 m 3 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-610-662 Supplier Restek™ Supplier No. 10258
Add to Cart
Add to Cart

General-Purpose Columns for Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners, Essential Oils, Semivolatiles

  • Removes all residual catalysts and low molecular weight fragments from the Rtx™-5 polymer
  • Provides a tight, unimodal distribution
  • Extremely low bleed
  • For Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles

Specifications

For Use With (Application) Drugs, Solvent Impurities, Pesticides, Hydrocarbons, PCB Congeners (e.g., Aroclor Mixes), Essential Oils, Semivolatiles
Film Thickness 1 μm
Length (Metric) 60 m
Polarity Low
Coating Fused Silica
Quantity 1/Pk.
USP Type G27, G36
Stationary Phase Rtx™-5
Diameter (Metric) Inner 0.53 mm
Max. Temperature (Metric) 330°C
Temperature Range (Metric) -60°C to +310/330°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.