Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RU 28318, potassium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective antagonist for the mineralocorticoid receptor (MR). Inhibits aldosterone production and secretion, and selectively decreases ex-vivo MR binding in the hippocampus of adrenalectomised rats. Also decreases blood pressure in female rats following central administration in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 440.66 |
| Percent Purity | >98% |
| Chemical Name or Material | RU 28318, potassium salt |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction