Learn More
Rubbermaid™ GroundsKeeper™ Smoking Management Receptacles
Catalog No. 19076524 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Color:
Black
Sandstone
Material:
Plastic, Galvanized Steel
Stainless Steel
Product Type:
Standard Receptacle
Tuscan Receptacle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Product Type
19-076-524 Sandstone Stainless Steel Tuscan Receptacle
19-076-522 Black Plastic, Galvanized Steel Standard Receptacle
19-076-523 Black Stainless Steel Tuscan Receptacle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-076-524 Supplier Rubbermaid™ Supplier No. FG9W300SSTON
Classic style, all-weather solution for unsightly cigarette waste
Standard Receptacle
  • Easy-to-clean, two-piece design
  • Constructed of smooth injection-molded plastic with inner galvanized-steel canister
  • Restricted opening shields cigarette waste from view, protects it from exposure to weather conditions, and limits flow of oxygen
  • Steel base plate adds weight for stability and discourages theft
Tuscan Receptacles
  • Provide upscale aesthetic appeal; blend with many architectural styles
  • Feature anchor holes for added security
  • Stainless-steel snuff plate minimizes smoldering
  • Choose from sandstone or black finish
Order Info
Dimensions: Standard receptacle, 12.25 x 12.25 x 39.44 in. (31.1 x 31.1 x 100cm); Tuscan receptacles, 13 x 13 x 38.38 in. (33 x 33 x 97.5cm). Shipping weight: Standard receptacle, 15.5 lb. (7kg); Tuscan receptacles, 15.4 lb. (7kg).
Specifications
Color Sandstone
Quantity 1 Ea.
Material Stainless Steel
Product Type Tuscan Receptacle