Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUBCNL Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB052401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052401 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052401 Supplier Novus Biologicals Supplier No. NBP298088F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUBCNL Polyclonal specifically detects RUBCNL in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RUBCNL
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias C13orf18, KIAA0226L, KIAA0226-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RUBCNL fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 80183
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.