Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RuBi-GABA
GREENER_CHOICESDP

Catalog No. 340010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-001-0 Supplier Tocris Bioscience™ Supplier No. 3400/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Caged GABA; excitable at visible wavelengths

Ruthenium-bipyridine-triphenylphosphine caged GABA (Cat. No. 0344) that is excited by visible wavelengths. Provides greater tissue penetration, less phototoxicity, faster photorelease kinetics and better spatial resolution than UV light-sensitive caged compounds. Produces GABA receptor-mediated currents in pyramidal neurons in vitro and displays no effect on endogenous GABAergic or glutamatergic transmission at concentrations effective for uncaging.

Chemical Identifiers

CAS 1028141-88-9
Molecular Formula C42H39F6N5O2P2Ru

Specifications

Quantity 10 mg
Formula Weight 922.8
Purity >98%
Molecular Weight (g/mol) 922.815
Chemical Name or Material RuBi-GABA
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.