Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium carbonate, 99.8% (metals basis)

Catalog No. p-7048445
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1056209 10 g
AA1056218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1056209 Supplier Thermo Scientific Chemicals Supplier No. 01056209
Only null left
Add to Cart

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol

Rubidium carbonate is used in glass-making by enhancing stability and durability as well as reducing its conductivity. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas. Rubidium carbonate glass has been extensively tested for use in anti-collision devices for motor vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate
PubChem CID 11431
IUPAC Name rubidium(1+);carbonate
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Powder
Quantity 10 g
Linear Formula Rb2CO3
Solubility Information Soluble in water.
Formula Weight 230.95
Percent Purity 99.8%
Odor Odorless
Sensitivity Very Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium carbonate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-530-9
RTECSNumber FG0650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.