Learn More
Rubidium carbonate, Puratronic™, 99.975% (metals basis)
Description
It finds it application as a raw material for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 837°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.975% (metals basis) |
Solubility Information | Soluble in water at 20°C 4500g/L. |
IUPAC Name | rubidium(1+);carbonate |
Formula Weight | 230.95 |
Odor | Odorless |
Sensitivity | Very Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 209-530-9 |
RTECSNumber | FG0650000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.