Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubisco Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11081553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11081553 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11081553 Supplier Novus Biologicals Supplier No. NB11081553
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Rubisco Polyclonal specifically detects Rubisco in Plant samples. It is validated for Western Blot.

Specifications

Antigen Rubisco
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias 5-bisphosphate carboxylase/oxygenase small subunit N-methyltransferase I [Spinacia oleracea], EC 2.1.1.43, rbcL, Ribulose-1
Host Species Chicken
Immunogen 1-491
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rubisco
Target Species Plant
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.