Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX1 and RUNX1T1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP259160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259160 Supplier Novus Biologicals Supplier No. NBP259160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RUNX1 and RUNX1T1 Polyclonal specifically detects RUNX1 and RUNX1T1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Chromatin Immunoprecipitation Sequencing.

Specifications

Antigen RUNX1 and RUNX1T1
Applications Western Blot, ELISA, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:500, Chromatin Immunoprecipitation (ChIP) 4 uL/IP, Chromatin Immunoprecipitation Sequencing
Formulation Serum with 0.05% Sodium Azide
Gene Alias Acute myeloid leukemia 1 protein, AML1, AML1acute myeloid leukemia 1, AML1-EVI-1, AML1T1protein CBFA2T1, CBFA2, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, Core-binding factor subunit alpha-2, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, EVI-1, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, PEBP2A2, Protein ETO, Protein MTG8, runt domain, alpha subunit 2, runt-related transcription factor 1, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22
Host Species Rabbit
Immunogen The exact immunogen is propietary information.
Purification Method Whole Antiserum
Quantity 100 μL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.