Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX2/CBFA1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224755SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224755SS 0.025 mg
NBP224755 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224755SS Supplier Novus Biologicals Supplier No. NBP224755SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

RUNX2/CBFA1 Polyclonal specifically detects RUNX2/CBFA1 in Human, Mouse, Chicken, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen RUNX2/CBFA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q13950
Gene Alias Acute myeloid leukemia 3 protein, AML3CCD, CBFA1MGC120022, Core-binding factor subunit alpha-1, MGC120023, PEA2-alpha A, PEBP2A, PEBP2A1CCD1, PEBP2aA1, PEBP2-alpha A, runt domain, alpha subunit 1, runt-related transcription factor 2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit
Gene Symbols RUNX2
Host Species Rabbit
Immunogen This RUNX2/CBFA1 Antibody was developed against a synthetic peptide corresponding to somewhere between amino acids 250-300 of human RUNX2 was used as immunogen for RUNX2/CBFA1 Antibody. RUNX2 and RUNX1 share an approximate 66% homology in peptide sequence used as immunogen.
Molecular Weight of Antigen 56.6 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 860
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, macaque, chimpanzee, orangutan, gorilla, cow, bison, horse, opossum and boar, and 90% homologous in dog and rat.
Target Species Human, Mouse, Chicken, Equine, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.