Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX2/CBFA1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$192.47 - $517.59

Specifications

Antigen RUNX2/CBFA1
Concentration 0.5 mg/mL
Dilution Western Blot 1 - 3 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224755SS
SDP
View Documents
Novus Biologicals
NBP224755SS
0.025 mg
Each for $192.47
Only null left
Add to Cart
 
NBP224755
SDP
View Documents
Novus Biologicals
NBP224755
0.1 mg
Each for $517.59
Only null left
Add to Cart
 
Description

Description

RUNX2/CBFA1 Polyclonal specifically detects RUNX2/CBFA1 in Human, Mouse, Chicken, Equine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

RUNX2/CBFA1
Western Blot 1 - 3 ug/mL
Polyclonal
Purified
RUO
Human, Mouse, Chicken, Equine, Primate
Q13950
860
This RUNX2/CBFA1 Antibody was developed against a synthetic peptide corresponding to somewhere between amino acids 250-300 of human RUNX2 was used as immunogen for RUNX2/CBFA1 Antibody. RUNX2 and RUNX1 share an approximate 66% homology in peptide sequence used as immunogen.
Primary
Store at -20C. Avoid freeze-thaw cycles.
56.6 kDa
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Transcription Factors and Regulators
PBS, 0.05% BSA with 0.05% Sodium Azide
Acute myeloid leukemia 3 protein, AML3CCD, CBFA1MGC120022, Core-binding factor subunit alpha-1, MGC120023, PEA2-alpha A, PEBP2A, PEBP2A1CCD1, PEBP2aA1, PEBP2-alpha A, runt domain, alpha subunit 1, runt-related transcription factor 2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit
RUNX2
IgG
Protein G purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, rhesus monkey, macaque, chimpanzee, orangutan, gorilla, cow, bison, horse, opossum and boar, and 90% homologous in dog and rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.