Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX3/CBFA3 Antibody (527327), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB303826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303826 Supplier Novus Biologicals Supplier No. FAB3765G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

RUNX3/CBFA3 Monoclonal antibody specifically detects RUNX3/CBFA3 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen RUNX3/CBFA3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 527327
Conjugate Alexa Fluor 488
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2
Host Species Mouse
Immunogen E. coli-derived recombinant human RUNX3/CBFA3, Lys186-Tyr415, Accession # Q13761
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 864
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.