Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUNX3/CBFA3 Antibody (K12P3F11*C9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250277AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250277AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250277AI Supplier Novus Biologicals Supplier No. NBP250277AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RUNX3/CBFA3 Monoclonal specifically detects RUNX3/CBFA3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen RUNX3/CBFA3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone K12P3F11*C9
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2
Gene Symbols RUNX3
Host Species Mouse
Immunogen Peptide - QFDRSFPTL
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 864
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.