Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruv B Antibody (RuvB 4G5/1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250279FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250279FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250279FR Supplier Novus Biologicals Supplier No. NBP250279FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ruv B Monoclonal antibody specifically detects Ruv B in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Ruv B
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone RuvB 4G5/1
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Ruv B protein from E. Coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.