Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL1 Rabbit anti-Human, DyLight 550, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB086977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086977 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086977 Supplier Novus Biologicals Supplier No. NBP300247R

Rabbit Polyclonal Antibody

RUVBL1 Polyclonal specifically detects RUVBL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RUVBL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias 49 kDa TATA box-binding protein-interacting protein, 54 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,49 kDa TBP-interacting protein, ECP54, INO80 complex subunit H, INO80HTIP49A, NMP 238, NMP238ECP-54, Nuclear matrix protein 238, PONTIN, Pontin 52, Pontin52, RuvB (E coli homolog)-like 1, ruvB-like 1, RuvB-like 1 (E. coli), RVB1, TATA binding protein interacting protein 49 kDa, TIH1, TIP49a, TIP49TAP54-alpha, TIP60-associated protein 54-alpha
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant RUVBL1 (Q9Y265-1; Met1-Lys456).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8607
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.