Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RUVBL2 Antibody (OTI2B9), HRP, Novus Biologicals™
SDP

Catalog No. NBP273960H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273960H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273960H Supplier Novus Biologicals Supplier No. NBP273960H

Mouse Monoclonal Antibody

RUVBL2 Monoclonal specifically detects RUVBL2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.

Specifications

Antigen RUVBL2
Applications Western Blot
Classification Monoclonal
Clone OTI2B9
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias 48 kDa TATA box-binding protein-interacting protein, 51 kDa erythrocyte cytosolic protein, EC 3.6.1, EC 3.6.4.12,48 kDa TBP-interacting protein, ECP51TIP49B, erythrocyte cytosolic protein, 51-KD, INO80 complex subunit J, INO80JTAP54-beta, Repressing pontin 52, REPTIN, Reptin 52, Reptin52, RuvB (E coli homolog)-like 2, ruvB-like 2, RuvB-like 2 (E. coli), RVB2, TBP-interacting protein, 48-KD, TIH2, TIP48ECP-51, TIP49b, TIP60-associated protein 54-beta
Gene Symbols RUVBL2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 113-370 of human RUVBL2 (NP_006657) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10856.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.