Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuvC Antibody (RuvC 5G9/3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP250280AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250280AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250280AG Supplier Novus Biologicals Supplier No. NBP250280AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RuvC Monoclonal antibody specifically detects RuvC in E. coli samples. It is validated for Western Blot, ELISA.

Specifications

Antigen RuvC
Applications Western Blot, ELISA
Classification Monoclonal
Clone RuvC 5G9/3
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Ruv C protein from E. Coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.