Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuvC Antibody (RuvC 7F1/5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309646 Supplier Novus Biologicals Supplier No. NBP250281MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RuvC Monoclonal antibody specifically detects RuvC in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen RuvC
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone RuvC 7F1/5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias crossover junction endodeoxyribonuclease RuvC, EC 3.1.22.4
Host Species Mouse
Immunogen Ruv C protein from E. Coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.