Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXFP3/RLN3R1/SALPR Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB045832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045832 Supplier Novus Biologicals Supplier No. NBP297628FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RXFP3/RLN3R1/SALPR Polyclonal specifically detects RXFP3/RLN3R1/SALPR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RXFP3/RLN3R1/SALPR
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor SALPR, GPCR135MGC141998, G-protein coupled receptor SALPR, relaxin 3 receptor 1, Relaxin family peptide receptor 3RLN3 receptor 1, relaxin/insulin-like family peptide receptor 3, relaxin-3 receptor 1, RLN3R1MGC142000, RXFPR3, SALPRG-protein coupled receptor GPCR135, somatostatin and angiotensin-like peptide receptor, Somatostatin- and angiotensin-like peptide receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RXFP3/RLN3R1/SALPR fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 51289
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.