Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rxi™-1HT Capillary Columns
SDP

Catalog No. 06611062 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
15 m
30 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
Film Thickness:
0.1 μm
0.25 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Film Thickness
06-611-062 30 m 0.25 mm 0.25 μm
06-611-060 15 m 0.25 mm 0.1 μm
06-611-061 30 m 0.25 mm 0.1 μm
06-611-063 15 m 0.32 mm 0.1 μm
06-611-064 30 m 0.32 mm 0.1 μm
06-611-065 30 m 0.32 mm 0.25 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-611-062 Supplier Restek™ Supplier No. 13952
Add to Cart
Add to Cart

Columns processed for high-temperature applications, such as high molecular weight hydrocarbons

  • Ultra-low bleed
  • Exceptional inertness
  • Reproducible performance, column after column

Specifications

For Use With (Application) High Molecular Weight Hydrocarbons, High-Temperature Applications
Film Thickness 0.25 μm
Length (Metric) 30 m
Material Fused Silica
Polarity Nonpolar
Coating Fused Silica
Quantity 1 Column
Stationary Phase Rxi™-1 HT
Diameter (Metric) Inner 0.25 mm
Temperature Range (Metric) -60°C to +400°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.