Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXR alpha/NR2B1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220130 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220130 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220130 Supplier Novus Biologicals Supplier No. NBP220130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RXR alpha/NR2B1 Polyclonal specifically detects RXR alpha/NR2B1 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen RXR alpha/NR2B1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.98mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunoprecipitation 1:100-1:500
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P19793
Gene Alias FLJ00280, FLJ00318, FLJ16020, MGC102720, NR2B1FLJ16733, Nuclear receptor subfamily 2 group B member 1, retinoic acid receptor RXR-alpha, Retinoid X receptor alpha, retinoid X receptor, alpha
Gene Symbols RXRA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Retinoid X Receptor alpha. The exact sequence is proprietary.
Molecular Weight of Antigen 51 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6256
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.