Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXR gamma/NR2B3 Antibody (PCRP-RXRG-5G6), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB187861 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187861 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187861 Supplier Novus Biologicals Supplier No. NBP314169V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RXR gamma/NR2B3 Monoclonal antibody specifically detects RXR gamma/NR2B3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen RXR gamma/NR2B3
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-RXRG-5G6
Conjugate DyLight 405
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias NR2B3retinoid X receptor-gamma, Nuclear receptor subfamily 2 group B member 3, retinoic acid receptor RXR-gamma, Retinoid X receptor gamma, retinoid X receptor, gamma, RXRC
Host Species Mouse
Immunogen Recombinant full-length human RXR gamma/NR2B3 protein (Uniprot: P48443)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6258
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.