Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RY586 Mouse Anti-Human CD83, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569651
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow cytometry
Specifications
| CD83 | |
| Monoclonal | |
| RY586 | |
| BL11; HB15; B-cell activation protein | |
| Human CD83 transfected COS cells | |
| 25 Tests | |
| 9308 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| HB15e | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction