Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RY586 Mouse Anti-Human CD83, BD Horizon™
Mouse Monoclonal Antibody
$190.50 - $447.50
Specifications
| Antigen | CD83 |
|---|---|
| Clone | HB15e |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RY586 |
Description
CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow cytometrySpecifications
| CD83 | |
| Flow Cytometry | |
| RY586 | |
| Mouse | |
| Human | |
| BL11; HB15; B-cell activation protein | |
| Human CD83 transfected COS cells | |
| Affinity Chromatography |
| HB15e | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 9308 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title