Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RY586 Mouse Anti-Human IL-22, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569661
Description
IL-22 Monoclonal antibody specifically detects IL-22 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications
IL-22 | |
Monoclonal | |
RY586 | |
interleukin-22; IL-10-related T-cell-derived inducible factor; IL-TIF; ILTIF; IL-22a, ilTIFa; Iltif; TIFa | |
Human IL-22 Recombinant Protein | |
25 Tests | |
50616, 50929 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Flow Cytometry | |
MH22B2 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction