Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY586 Mouse Anti-Human IL-22, BD Horizon™

Mouse Monoclonal Antibody

$194.00 - $448.00

Specifications

Antigen IL-22
Clone MH22B2
Applications Flow Cytometry
Classification Monoclonal
Conjugate RY586
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB569660
View Documents
BD Biosciences
569660
100 Tests
Each for $448.00
Only null left
Add to Cart
 
BDB569661
View Documents
BD Biosciences
569661
25 Tests
Each for $194.00
Only null left
Add to Cart
 
Description

Description

IL-22 Monoclonal antibody specifically detects IL-22 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications

Specifications

IL-22
Flow Cytometry
RY586
Mouse
Human, Mouse
interleukin-22; IL-10-related T-cell-derived inducible factor; IL-TIF; ILTIF; IL-22a, ilTIFa; Iltif; TIFa
Human IL-22 Recombinant Protein
Affinity Chromatography
MH22B2
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
50616, 50929
IgG2a κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.