Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine Receptor 2 Antibody (C3-33), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB013285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013285 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013285 Supplier Novus Biologicals Supplier No. NBP280143G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ryanodine Receptor 2 Monoclonal specifically detects Ryanodine Receptor 2 in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Ryanodine Receptor 2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C3-33
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias arrhythmogenic right ventricular dysplasia 2, ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, Cardiac muscle-type ryanodine receptor, cardiac-type ryanodine receptor, hRYR-2, islet-type ryanodine receptor, kidney-type ryanodine receptor, ryanodine receptor 2, ryanodine receptor 2 (cardiac), RyR2, RYR-2, type 2 ryanodine receptor, VTSIP
Gene Symbols RYR2
Host Species Mouse
Immunogen Canine cardiac ryanodine receptor (RyR2)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6262
Test Specificity Detects Ryanodine Receptor 2. Also shows cross-reactivity with Ryanodine Receptor 1 in amphibian, canine, chicken, fish, guinea pig and rat tissues.
Target Species Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Fish, Guinea Pig, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.