Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine Receptor Antibody (34C), FITC, Novus Biologicals™
SDP

Catalog No. NB004028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004028 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004028 Supplier Novus Biologicals Supplier No. NB300543F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ryanodine Receptor Monoclonal specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Ryanodine Receptor
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 34C
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, cardiac-type ryanodine receptor, CCO, central core disease of muscle, islet-type ryanodine receptor, kidney-type ryanodine receptor, MHS, MHS1, PPP1R137, protein phosphatase 1, regulatory subunit 137, ryanodine receptor 1, ryanodine receptor 1 (skeletal), ryanodine receptor 2, ryanodine receptor 2 (cardiac), RYDR, RYR, RYR-1, RYR-2, sarcoplasmic reticulum calcium release channel, Skeletal muscle calcium release channel, skeletal muscle ryanodine receptor, SKRR, type 1-like ryanodine receptor, type 2 ryanodine receptor, VTSIP
Gene Symbols RYR1
Host Species Mouse
Immunogen Partially purified chicken pectoral muscle ryanodine receptor.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This antibody detects ryanodine receptor Ryanodine Receptor-1 and Ryanodine Receptor-2 isoforms in amphibian, chicken, bovine, canine, fish, human, non-human primate, mouse, rabbit, mink, rat and sheep tissues. In chicken, it detects the alpha, beta and cardiac isoforms. This antibody also detects Ryanodine Receptor-3 in mouse cells. In frog, it detects the alpha and beta isoforms and in fish it detects the alpha isoform.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.