Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryanodine Receptor Antibody (34C), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359417 Supplier Novus Biologicals Supplier No. NB300543MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ryanodine Receptor Monoclonal antibody specifically detects Ryanodine Receptor in Human, Mouse, Rat, Amphibian, Bovine, Canine, Fish, Ferret, Mink, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Neutralization

Specifications

Antigen Ryanodine Receptor
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Blocking Assay
Classification Monoclonal
Clone 34C
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias ARVC2, ARVD2, cardiac muscle ryanodine receptor-calcium release channel, cardiac-type ryanodine receptor, CCO, central core disease of muscle, islet-type ryanodine receptor, kidney-type ryanodine receptor, MHS, MHS1, PPP1R137, protein phosphatase 1, regulatory subunit 137, ryanodine receptor 1, ryanodine receptor 1 (skeletal), ryanodine receptor 2, ryanodine receptor 2 (cardiac), RYDR, RYR, RYR-1, RYR-2, sarcoplasmic reticulum calcium release channel, Skeletal muscle calcium release channel, skeletal muscle ryanodine receptor, SKRR, type 1-like ryanodine receptor, type 2 ryanodine receptor, VTSIP
Host Species Mouse
Immunogen Partially purified chicken pectoral muscle ryanodine receptor.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6261
Target Species Human, Mouse, Rat, Amphibian, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.