Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RYBP/DEDAF Antibody (OTI2B4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273962FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273962FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273962FR Supplier Novus Biologicals Supplier No. NBP273962FR

Mouse Monoclonal Antibody

RYBP/DEDAF Monoclonal specifically detects RYBP/DEDAF in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot.

Specifications

Antigen RYBP/DEDAF
Applications Western Blot
Classification Monoclonal
Clone OTI2B4
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias AAP1, APAP-1, Apoptin-associating protein 1, Death effector domain-associated factor, DEDAFYY1 and E4TF1-associated factor 1, RING1 and YY1 binding protein, RING1 and YY1-binding protein, ring1 interactor RYBP, YEAF1DED-associated factor, YY1 and E4TF1 associated factor 1
Gene Symbols RYBP
Host Species Mouse
Immunogen Full length recombinant protein of human RYBP/DEDAF (NP_036366) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 23429.0
Target Species Human, Mouse, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.