Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ryk Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB324879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324879 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324879 Supplier Novus Biologicals Supplier No. NBP299540APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ryk Polyclonal antibody specifically detects Ryk in Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Ryk
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias D3S3195, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, JTK5, JTK5A, JTK5A protein tyrosine kinase, RYK receptor-like tyrosine kinase, RYK1, tyrosine-protein kinase RYK
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ryk
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6259
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.