Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Ryuvidine

Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of SETD8 protein lysine methyltransferase (PKMT) (IC50 = 0.5 μM); suppresses H4K20 monomethylation in vitro. Also inhibits cyclin-dependent kinase (CDK) 4 (IC50 = 6.0 μM at CDK4/cyclin D1). Induces S phase accumulation in HEK293T cells. Cytotoxic against a range of human cancer cells.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 284.33 |
Purity | >98% |
Molecular Weight (g/mol) | 284.333 |
Chemical Name or Material | Ryuvidine |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction